Novus Biologicals
Manufacturer Code:NBP297021AF488
Catalog # NB080678
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with purified recombinant RBP5 (P82980 Met1-Arg135). |
Conjugate | Alexa Fluor 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cellular retinol-binding protein III; Cellular retinol-binding protein III CRBP3 CRBPIII CRBP-III HRBPiso putative cellular retinol-binding protein CRBP III retinol binding protein 5 cellular retinol-binding protein 5 retinol-binding protein 5 cellular; CRBP-III; HRBPiso; putative cellular retinol-binding protein CRBP III; retinol binding protein 5, cellular; Retinol-binding protein 5; retinol-binding protein 5, cellular
Gene Aliases: CRBP-III; CRBP3; CRBPIII; HRBPiso; RBP5
UniProt ID: (Human) P82980
Entrez Gene ID: (Human) 83758
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.