R&D Systems
Manufacturer Code:MAB34761
Catalog # MAB34761
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 423619 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant mouse RBP4/Retinol-Binding Protein 4 Met1-Leu201 Accession # NP_035385 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: interstitial Plasma retinol-binding protein retinol binding protein 4 plasma RetinolBinding Protein 4 retinol-binding protein 4 plasma; Plasma retinol-binding protein; Plasma retinol-binding protein(1-176); Plasma retinol-binding protein(1-179); Plasma retinol-binding protein(1-181); Plasma retinol-binding protein(1-182); PRBP; RBP; retinol binding protein 4, plasma; Retinol-binding protein 4; retinol-binding protein 4, interstitial
Gene Aliases: MCOPCB10; PRO2222; RBP4; RDCCAS
UniProt ID: (Human) P02753
Entrez Gene ID: (Human) 5950
Molecular Function: transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.