Invitrogen
Manufacturer Code:MA515838
Catalog # PIMA515838
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 4C2 |
Immunogen | Purified recombit fragment of human RBP expressed in E. Coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Plasma retinol-binding protein; Plasma retinol-binding protein(1-176); Plasma retinol-binding protein(1-179); Plasma retinol-binding protein(1-181); Plasma retinol-binding protein(1-182); PRBP; PRO2222 PRBP RBP plasma retinol-binding protein retinol-binding protein 4 retinol-binding protein 4 interstitial; RBP; retinol binding protein 4, plasma; Retinol-binding protein 4; retinol-binding protein 4, interstitial
Gene Aliases: MCOPCB10; PRO2222; RBP4; RDCCAS
UniProt ID: (Human) P02753
Entrez Gene ID: (Human) 5950
Molecular Function: transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.