Invitrogen
Manufacturer Code:PA579919
Catalog # PIPA579919
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived mouse RBP4 recombit protein (Position: E19-L201). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Plasma retinol-binding protein; Plasma retinol-binding protein(1-176); Plasma retinol-binding protein(1-179); Plasma retinol-binding protein(1-181); Plasma retinol-binding protein(1-182); PRBP; RBP; retinol binding protein 4, cellular; retinol binding protein 4, plasma; Retinol-binding protein 4; Retinol-binding protein 4 Plasma retinol-binding protein PRBP RBP Rbp4; retinol-binding protein 4, interstitial
Gene Aliases: MCOPCB10; PRO2222; Rbp-4; RBP4; RBPA; RDCCAS
UniProt ID: (Human) P02753, (Rat) P04916, (Mouse) Q00724
Entrez Gene ID: (Human) 5950, (Rat) 25703, (Mouse) 19662
Molecular Function:
transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.