Invitrogen
Manufacturer Code:PA518632
Catalog # PIPA518632
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (QKGNDDHWIVDTDYD) corresponding to the internal amino acids of RBP4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Plasma retinol-binding protein; Plasma retinol-binding protein(1-176); Plasma retinol-binding protein(1-179); Plasma retinol-binding protein(1-181); Plasma retinol-binding protein(1-182); PRBP; RBP; RBP4 retinol binding protein 4 plasma retinol-binding protein 4 interstitial retinol-binding protein 4 plasma; retinol binding protein 4, plasma; Retinol-binding protein 4; retinol-binding protein 4, interstitial
Gene Aliases: MCOPCB10; PRO2222; RBP4; RDCCAS
UniProt ID: (Human) P02753
Entrez Gene ID: (Human) 5950
Molecular Function: transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.