Invitrogen
Manufacturer Code:PA588586
Catalog # PIPA588586
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 1-134 of human RBP2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cellular retinol-binding protein II; CRBP-II; retinol binding protein 2, cellular; Retinol-binding protein 2; retinol-binding protein 2, cellular
Gene Aliases: CRABP-II; Crbp-2; CRBP2; CRBPII; Rbp-2; RBP2; RBPC2
UniProt ID: (Human) P50120, (Mouse) Q08652
Entrez Gene ID: (Human) 5948, (Mouse) 19660
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.