Invitrogen
Manufacturer Code:PA557438
Catalog # PIPA557438
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human RBP2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cellular retinol-binding protein II; CRABP-II CRBP2 CRBPII RBPC2 CRBP-II cellular retinol-binding protein II retinol-binding protein 2 retinol-binding protein 2 cellular; CRBP-II; retinol binding protein 2, cellular; Retinol-binding protein 2; retinol-binding protein 2, cellular
Gene Aliases: CRABP-II; CRBP2; CRBPII; RBP2; RBPC2
UniProt ID: (Human) P50120
Entrez Gene ID: (Human) 5948
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.