Novus Biologicals
Manufacturer Code:NBP297451FR
Catalog # NB043635
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human RBP2 fragment. |
Conjugate | DyLight 680 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cellular Cellular retinol-binding protein II CRABP-II CRBP2CRBP-II retinol binding protein 2 cellular; Cellular retinol-binding protein II; CRBP-II; retinol binding protein 2, cellular; Retinol-binding protein 2; retinol-binding protein 2, cellular
Gene Aliases: CRABP-II; CRBP2; CRBPII; RBP2; RBPC2
UniProt ID: (Human) P50120
Entrez Gene ID: (Human) 5948
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.