Invitrogen
Manufacturer Code:PA5110725
Catalog # PIPA5110725
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Protein corresonding to Human RBP1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C CRABP-I Cellular retinol-binding protein Cellular retinol-binding protein I CRABP-I CRBP CRBP1 Crbpi CRBP-I RBP RBP1 Rbp-1 RBPC Retinol-binding protein 1; cellular retinol binding protein 1; Cellular retinol-binding protein; Cellular retinol-binding protein I; CRBP; CRBP-I; Retinol-binding protein 1; retinol-binding protein 1, cellular
Gene Aliases: CRABP-I; CRBP; CRBP1; CRBPI; RBP1; RBPC
UniProt ID: (Human) P09455
Entrez Gene ID: (Human) 5947
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.