Invitrogen
Manufacturer Code:PA1727
Catalog # APPG27
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide: D(47) H M I I R T L S T F R N Y I M D F Q(65) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cellular retinol-binding protein; Cellular retinol-binding protein CRBP CRBP1 RBP1 RBP-1; Cellular retinol-binding protein I; CRBP; CRBP-I; Retinol-binding protein 1
Gene Aliases: Rbp-1; Rbp1
UniProt ID: (Rat) P02696
Entrez Gene ID: (Rat) 25056
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.