Proteintech
Manufacturer Code:226831AP150UL
Catalog # 50-173-3922
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | RBP1 Fusion Protein Ag18572 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cellular retinol binding protein 1; Cellular retinol-binding protein; Cellular retinol-binding protein I; CRABP I CRBP CRBP I CRBP1 CRBPI RBP1 RBPC Retinol binding protein 1; CRBP; CRBP-I; mCRBPI; Retinol-binding protein 1; retinol-binding protein 1, cellular
Gene Aliases: CRABP-I; CRBP; CRBP1; CRBPI; Rbp-1; RBP1; RBPC
UniProt ID: (Human) P09455, (Mouse) Q00915, (Rat) P02696
Entrez Gene ID: (Human) 5947, (Mouse) 19659, (Rat) 25056
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.