Proteintech
Manufacturer Code:179941AP150UL
Catalog # 50-173-1923
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | RBMX2 Fusion Protein Ag12439 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CGI 79 RBMX2; RNA-binding motif protein, X-linked 2
Gene Aliases: 2700089A17Rik; 2810411A03Rik; CGI-79; RBMX2; RGD1562693; Snu17
UniProt ID: (Human) Q9Y388, (Rat) B0BN49, (Mouse) Q8R0F5
Entrez Gene ID: (Human) 51634, (Rat) 367930, (Mouse) 209003
Molecular Function:
RNA binding protein
mRNA processing factor
mRNA splicing factor
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.