OriGene
Manufacturer Code:TA808580
Catalog # 50-168-2235
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2H1 |
Immunogen | Full length human recombit protein of human RBMS1 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: c-myc gene single strand binding protein 2; C2orf12 HCC-4 MSSP MSSP-1 MSSP-2 MSSP-3 SCR2 YC1; cervical cancer oncogene 4; RNA binding motif, single stranded interacting protein 1; RNA-binding motif, single-stranded-interacting protein 1; Single-stranded DNA-binding protein MSSP-1; suppressor of cdc 2 (cdc13) with RNA binding motif 2; Suppressor of CDC2 with RNA-binding motif 2
Gene Aliases: C2orf12; HCC-4; MSSP; MSSP-1; MSSP-2; MSSP-3; MSSP1; RBMS1; SCR2; YC1
UniProt ID: (Human) Q53P46
Entrez Gene ID: (Human) 5937
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.