Invitrogen
Manufacturer Code:PA531887
Catalog # PIPA531887
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein fragment corresponding to a region within amino acids 1 and 206 of Human RBM4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hLark; Lark homolog; LARK RBM4A ZCCHC21 ZCRB3A RNA-binding motif protein 4a RNA-binding protein 4 lark homolog transcriptional coactivator CoAZ zinc finger CCHC-type and RNA binding motif 3A; mLark; RNA-binding motif protein 4; RNA-binding motif protein 4a; RNA-binding protein 4; transcriptional coactivator CoAZ; zinc finger CCHC-type and RNA binding motif 3A
Gene Aliases: 4921506I22Rik; LARK; Lark1; Mlark; RBM4; RBM4A; ZCCHC21; ZCRB3A
UniProt ID: (Human) Q9BWF3, (Mouse) Q8C7Q4
Entrez Gene ID: (Human) 5936, (Mouse) 19653
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.