Invitrogen
Manufacturer Code:PA5100075
Catalog # PIPA5100075
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human RBM4 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 4921506I22Rik hLark LARK Lark homolog Lark1 Mlark RBM4 RBM4A RNA binding motif protein 4 rna-binding RNA-binding motif protein 4 RNA-binding motif protein 4a RNA-binding protein 4 transcriptional coactivator CoAZ ZCCHC21 ZCRB3A zinc finger CCHC-type and RNA binding motif 3A; hLark; Lark homolog; mLark; RNA binding motif protein 4; RNA-binding motif protein 4; RNA-binding motif protein 4a; RNA-binding protein 4; transcriptional coactivator CoAZ; zinc finger CCHC-type and RNA binding motif 3A
Gene Aliases: 4921506I22Rik; LARK; Lark1; Mlark; RBM4; RBM4A; ZCCHC21; ZCRB3A
UniProt ID: (Human) Q9BWF3, (Mouse) Q8C7Q4
Entrez Gene ID: (Human) 5936, (Mouse) 19653, (Rat) 293663
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.