Novus Biologicals
Manufacturer Code:NBP220106
Catalog # NBP220106
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human RBM4. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DKFZp547K0918 FLJ36727 hLark Lark homolog MGC75138 RBM4. LARK RBM4ALARK RNA binding motif protein 4 RNA-binding motif protein 4 RNA-binding motif protein 4a RNA-binding protein 4 transcriptional coactivator CoAZ ZCCHC21 ZCRB3A zinc finger CCHC-type and RNA binding motif 3A; hLark; Lark homolog; RNA-binding motif protein 4; RNA-binding motif protein 4a; RNA-binding protein 4; transcriptional coactivator CoAZ; zinc finger CCHC-type and RNA binding motif 3A
Gene Aliases: LARK; RBM4; RBM4A; ZCCHC21; ZCRB3A
UniProt ID: (Human) Q9BWF3
Entrez Gene ID: (Human) 5936
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.