Invitrogen
Manufacturer Code:PA541952
Catalog # PIPA541952
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human RBM24 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: RNA binding protein; RNA-binding motif protein 24; RNA-binding protein 24; RNA-binding protein SEB-4; RNA-binding region (RNP1, RRM) containing 6; RNA-binding region-containing protein 6
Gene Aliases: 6330546B05Rik; AI606861; dJ259A10.1; RBM24; RNPC6; seb-4
UniProt ID: (Human) Q9BX46, (Mouse) D3Z4I3, (Zebrafish) Q76LC6
Entrez Gene ID: (Bovine) 524220, (Human) 221662, (Dog) 610452, (Horse) 100051943, (Guinea pig) 100720427, (Mouse) 666794, (Rat) 690139, (Zebrafish) 405801
Molecular Function:
ribosomal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.