Novus Biologicals
Manufacturer Code:NBP257328
Catalog # NBP257328
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:PALIQRPFGIPAHYVYPQAFVQPGVVIPHVQPTA |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: dJ259A10.1 FLJ26355 FLJ30829 FLJ37697 RNA binding motif protein 24 RNA-binding motif protein 24 RNA-binding protein 24 RNA-binding region (RNP1 RRM) containing 6 RNA-binding region-containing protein 6 RNPC6; RNA-binding motif protein 24; RNA-binding protein 24; RNA-binding region (RNP1, RRM) containing 6; RNA-binding region-containing protein 6
Gene Aliases: dJ259A10.1; RBM24; RNPC6
UniProt ID: (Human) Q9BX46
Entrez Gene ID: (Human) 221662
Molecular Function: ribosomal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.