Proteintech
Manufacturer Code:181781AP150UL
Catalog # 50-173-2033
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | RBM24 Fusion Protein Ag12871 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: RBM24 RNA binding motif protein 24 RNA binding protein 24 RNPC6; RNA-binding motif protein 24; RNA-binding protein 24; RNA-binding region (RNP1, RRM) containing 6; RNA-binding region-containing protein 6
Gene Aliases: 6330546B05Rik; AI606861; dJ259A10.1; RBM24; RNPC6
UniProt ID: (Human) Q9BX46, (Mouse) D3Z4I3
Entrez Gene ID: (Human) 221662, (Mouse) 666794, (Rat) 690139
Molecular Function: ribosomal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.