Proteintech
Manufacturer Code:105871AP150UL
Catalog # 50-172-6428
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | RBM15 Fusion Protein Ag0938 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: one twenty two protein; One twenty two protein 1 OTT OTT1 RBM15 RNA binding motif protein 15 SPEN; one twenty-two; One-twenty two protein 1; putative RNA-binding protein 15; RNA binding motif protein 15; RNA-binding motif protein 15; RNA-binding protein 15
Gene Aliases: C230088J01Rik; Kiaa4257; mKIAA1438; OTT; OTT1; RBM15; SPEN
UniProt ID: (Human) Q96T37, (Mouse) Q0VBL3
Entrez Gene ID: (Human) 64783, (Rat) 684233, (Mouse) 229700
Molecular Function:
RNA binding protein
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.