Novus Biologicals
Manufacturer Code:NBP192693SS
Catalog # NBP192693SS
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1B7 |
Immunogen | N-terminal 99 amino acids of human FOX3 as expressed in and purified from E. coli |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: FLJ56884 FLJ58356 Fox-1 homolog C FOX3 FOX-3 hexaribonucleotide binding protein 3 HRNBP3 NeuN NeuN flow cytometry neuronal nuclei RBFOX3 RNA binding protein fox-1 homolog 3 RNA binding protein fox-1 homolog (C. elegans) 3; Fox-1 homolog C; hexaribonucleotide binding protein 3; NeuN antigen; neuronal nuclei; Neuronal nuclei antigen; RNA binding protein fox-1 homolog 3; RNA binding protein, fox-1 homolog (C. elegans) 3
Gene Aliases: FOX-3; FOX3; HRNBP3; NEUN; RBFOX3
UniProt ID: (Human) A6NFN3
Entrez Gene ID: (Human) 146713
Molecular Function:
RNA binding protein
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.