Invitrogen
Manufacturer Code:PA540958
Catalog # PIPA540958
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human RBBP9 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: B5T overexpressed gene protein; B5T-overexpressed gene protein; Protein BOG; Putative hydrolase RBBP9; RBBP-10; RBBP-9; retinoblastoma binding protein 9; Retinoblastoma binding protein 9 BOG MGC9236 RBBP10; Retinoblastoma-binding protein 10; Retinoblastoma-binding protein 9; retinoma-binding protein 9; Serine hydrolase RBBP9
Gene Aliases: BOG; im:7247963; RBBP10; RBBP9; si:ch211-258l4.7; zgc:152807
UniProt ID: (Human) O75884, (Rat) O88350, (Mouse) O88851
Entrez Gene ID: (Human) 10741, (Bovine) 513835, (Horse) 100050518, (Dog) 607819, (Rabbit) 100346098, (Rat) 29459, (Mouse) 26450, (Guinea pig) 100722474, (Zebrafish) 553083
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.