Abnova
Manufacturer Code:PAB17317
Catalog # 89-118-976
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to internal of human RBBP8. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of RBBP8.
For Research Use Only
Protein Aliases: CTBP-interacting protein; CTIP; DNA endonuclease RBBP8; RBBP-8; retinoblastoma binding protein 8; Retinoblastoma-binding protein 8; Retinoblastoma-interacting protein and myosin-like; RIM; SAE2; Sporulation in the absence of SPO11 protein 2 homolog
Gene Aliases: COM1; CTIP; JWDS; RBBP8; RIM; SAE2; SCKL2
UniProt ID: (Human) Q99708
Entrez Gene ID: (Human) 5932
Molecular Function:
transcription cofactor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.