Proteintech
Manufacturer Code:100691AP150UL
Catalog # 50-172-5998
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | RB1CC1 Fusion Protein Ag0003 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 200 kDa FAK family kinase-interacting protein; CC1 DRAGOU14 FIP200 KIAA0203 RB1 inducible coiled coil 1 RB1CC1 RBICC; coiled coil forming protein 1; Coiled-coil-forming protein 1; FAK family kinase-interacting protein of 200 kDa; FIP200; LaXp180; phosphatase 1, regulatory subunit 131; RB1-inducible coiled-coil 1; RB1-inducible coiled-coil protein 1
Gene Aliases: 2900055E04Rik; 5930404L04Rik; ATG17; CC1; FIP200; KIAA0203; LaXp180; PPP1R131; RB1CC1; RBICC
UniProt ID: (Human) Q8TDY2, (Mouse) Q9ESK9
Entrez Gene ID: (Human) 9821, (Mouse) 12421
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.