Abnova
Manufacturer Code:H00009821K
Catalog # 89-203-224
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human RB1CC1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human RB1CC1 peptide using ARM Technology.
For Research Use Only
Protein Aliases: 200 kDa FAK family kinase-interacting protein; CC1 DRAGOU14 FIP200; FAK family kinase-interacting protein of 200 kDa; FIP200; phosphatase 1, regulatory subunit 131; RB1-inducible coiled-coil 1; RB1-inducible coiled-coil protein 1
Gene Aliases: ATG17; CC1; FIP200; KIAA0203; PPP1R131; RB1CC1; RBICC
UniProt ID: (Human) Q8TDY2
Entrez Gene ID: (Human) 9821
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.