Novus Biologicals
Manufacturer Code:NBP273808AF488
Catalog # NB273808A48
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1F7 |
Immunogen | Full length recombinant protein of human RASD2 (NP_055125) produced in HEK293T cells. |
Conjugate | Alexa Fluor 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: GTP-binding protein Rhes; GTP-binding protein Rhes MGC:4834 RASD family member 2 Rhes TEM2Ras homolog enriched in striatum Tumor endothelial marker 2; Ras homolog enriched in striatum; RASD family, member 2; Tumor endothelial marker 2
Gene Aliases: MGC:4834; RASD2; Rhes; TEM2
UniProt ID: (Human) Q96D21
Entrez Gene ID: (Human) 23551
Molecular Function: G-protein enzyme modulator small GTPase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.