Invitrogen
Manufacturer Code:PA520438
Catalog # PIPA520438
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A 15 amino acid peptide near the carboxy terminus of the human TEM2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GTP-binding protein Rhes; MGC:4834 Rhes TEM2 GTP-binding protein Rhes Ras homolog enriched in striatum tumor endothelial marker 2; Ras homolog enriched in striatum; RASD family, member 2; SE6C; Tumor endothelial marker 2
Gene Aliases: 4930526B11Rik; AU045414; MGC:4834; RASD2; Rhes; TEM-2; TEM2
UniProt ID: (Human) Q96D21, (Rat) P63033, (Mouse) P63032
Entrez Gene ID: (Human) 23551, (Rat) 171099, (Mouse) 75141
Molecular Function: G-protein enzyme modulator small GTPase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.