Abnova
Manufacturer Code:PAB13306
Catalog # 89-116-555
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to N-terminus 16 amino acids of human RASD2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C for three months. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of RASD2.
For Research Use Only
Protein Aliases: GTP-binding protein Rhes; MGC:4834; Ras homolog enriched in striatum; RASD family, member 2; Rhes; TEM2; Tumor endothelial marker 2
Gene Aliases: MGC:4834; RASD2; Rhes; TEM2
UniProt ID: (Human) Q96D21
Entrez Gene ID: (Human) 23551
Molecular Function:
G-protein
enzyme modulator
small GTPase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.