Invitrogen
Manufacturer Code:PA572536
Catalog # PIPA572536
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 369-399 amino acids from the C-terminal region of human RARS2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Arginyl-tRNA synthetase; ArgRS; Probable arginine--tRNA ligase mitochondrial Arginyl-tRNA synthetase ArgRS RARS2 RARSL; Probable arginine--tRNA ligase, mitochondrial; probable arginyl-tRNA synthetase, mitochondrial
Gene Aliases: ArgRS; DALRD2; PCH6; PRO1992; RARS2; RARSL
UniProt ID: (Human) Q5T160
Entrez Gene ID: (Human) 57038
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.