Proteintech
Manufacturer Code:668151IG150UL
Catalog # 50-173-7206
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1C8G4 |
Immunogen | RARS Fusion Protein Ag26399 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: arginine tRNA ligase 1, cytoplasmic; Arginine tRNA ligase arginyl tRNA synthetase ArgRS DALRD1 RARS; Arginine--tRNA ligase, cytoplasmic; Arginyl-tRNA synthetase; arginyl-tRNA synthetase, cytoplasmic; ArgRS
Gene Aliases: 2610011N19Rik; 2610037E21Rik; AL033339; ArgRS; AW985894; DALRD1; HLD9; RARS; RARS1
UniProt ID: (Human) P54136, (Rat) P40329, (Mouse) Q9D0I9
Entrez Gene ID: (Human) 5917, (Rat) 287191, (Mouse) 104458
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.