Proteintech
Manufacturer Code:114241AP150UL
Catalog # 50-172-7148
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | RARG Fusion Protein Ag1991 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: NR1B3 RAR gamma RARC RARG Retinoic acid receptor gamma retinoic acid receptor gamma; Nuclear receptor subfamily 1 group B member 3; RAR gamma 2; RAR-gamma; retinoic acid nuclear receptor gamma variant 1; retinoic acid nuclear receptor gamma variant 2; Retinoic acid receptor gamma; retinoic acid receptor gamma 4; retinoic acid receptor, gamma
Gene Aliases: NR1B3; RAR-gamma; RARC; RARD; RARG; RARgamma2
UniProt ID: (Human) J3KNP6, (Mouse) P18911
Entrez Gene ID: (Human) 5916, (Mouse) 19411, (Rat) 685072
Molecular Function:
nuclear hormone receptor
nucleic acid binding
receptor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.