Novus Biologicals
Manufacturer Code:NBP247314
Catalog # NBP247314
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to amino acids: MATNKERLFAAGALGPGSGYPGAGFPFAFPGALRGSPPFEMLSPSFRGLGQPDLPKEMASLSVET |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: NR1B3RARC Nuclear receptor subfamily 1 group B member 3 RAR-gamma retinoic acid receptor gamma retinoic acid receptor gamma; Nuclear receptor subfamily 1 group B member 3; RAR-gamma; retinoic acid nuclear receptor gamma variant 1; retinoic acid nuclear receptor gamma variant 2; Retinoic acid receptor gamma; retinoic acid receptor, gamma
Gene Aliases: NR1B3; RARC; RARG
UniProt ID: (Human) J3KNP6
Entrez Gene ID: (Human) 5916
Molecular Function:
nuclear hormone receptor
nucleic acid binding
receptor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.