Novus Biologicals
Manufacturer Code:NBP273802JF549
Catalog # N273802J549
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2C3 |
Immunogen | Recombinant protein fragment corresponding to amino acids 52-299 of human RAR alpha/NR1B1 (NP_000955) produced in E.coli. |
Conjugate | Janelia Fluor 549 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: alpha polypeptide RAR-alpha retinoic acid receptor alpha; Nuclear receptor subfamily 1 group B member 1; nucleophosmin-retinoic acid receptor alpha fusion protein NPM-RAR long form; RAR-alpha; retinoic acid nuclear receptor alpha variant 1; retinoic acid nuclear receptor alpha variant 2; Retinoic acid receptor alpha; retinoic acid receptor, alpha polypeptide
Gene Aliases: NR1B1; RAR; RARA
UniProt ID: (Human) P10276
Entrez Gene ID: (Human) 5914
Molecular Function: nuclear hormone receptor nucleic acid binding receptor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.