Invitrogen
Manufacturer Code:PA542285
Catalog # PIPA542285
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human RAPSN |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 43 kD receptor-associated protein of the synapse; 43 kDa postsynaptic protein; 43 kDa receptor-associated protein of the synapse; 43kDa acetylcholine receptor-associated protein; Acetylcholine receptor-associated 43 kDa protein; RAPsyn; receptor-associated protein of the synapse; Receptor-associated protein of the synapse CMS1D CMS1E MGC3597 RAPSYN RNF205; receptor-associated protein of the synapse, 43 kDa; RING finger protein 205; twitch once; two
Gene Aliases: 43kDa; CMS11; CMS4C; FADS; Nraps; Raps; RAPSN; RAPSYN; RNF205
UniProt ID: (Human) Q13702, (Mouse) P12672
Entrez Gene ID: (Rabbit) 100354253, (Horse) 100051540, (Human) 5913, (Dog) 609725, (Bovine) 541061, (Rat) 362161, (Guinea pig) 100729613, (Mouse) 19400, (Zebrafish) 352942
Molecular Function:
G-protein modulator
enzyme modulator
guanyl-nucleotide exchange factor
transmembrane receptor regulatory/adaptor protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.