Invitrogen
Manufacturer Code:PA5115824
Catalog # PIPA5115824
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human Rapsyn corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 43 kDa postsynaptic protein; 43 kda postsynaptic protein 43 kDa receptor-associated protein of the synapse 43kDa 43kDa acetylcholine receptor-associated protein acetylcholine receptor-associated 43 kda protein CMS11 CMS1D CMS1E CMS4C FADS MGC3597 Nraps Raps RAPSN rapsyn receptor associated protein of the synapse receptor-associated protein of the synapse receptor-associated protein of the synapse 43 kDa receptor-associated protein of the synapse 43kD RING finger protein 205 RNF205; 43 kDa receptor-associated protein of the synapse; 43kDa acetylcholine receptor-associated protein; Acetylcholine receptor-associated 43 kDa protein; RAPsyn; receptor-associated protein of the synapse; receptor-associated protein of the synapse, 43 kDa; RING finger protein 205
Gene Aliases: 43kDa; CMS11; CMS4C; FADS; Nraps; Raps; RAPSN; RAPSYN; RNF205
UniProt ID: (Human) Q13702, (Mouse) P12672
Entrez Gene ID: (Human) 5913, (Rat) 362161, (Mouse) 19400
Molecular Function: G-protein modulator enzyme modulator guanyl-nucleotide exchange factor transmembrane receptor regulatory/adaptor protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.