Invitrogen
Manufacturer Code:MA535451
Catalog # PIMA535451
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC0908 |
Immunogen | A synthesized peptide derived from human Epac1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: 2310016P22Rik 9330170P05Rik bcm910 BOS_5155 cAMP-GEFI cAMP-regulated guanine nucleotide exchange factor I CGEF1 Epac EPAC 1 EPAC1 Exchange factor directly activated by cAMP 1 Exchange protein directly activated by cAMP 1 HSU79275 MGC21410 Rap guanine nucleotide exchange factor 3 Rap1 guanine-nucleotide-exchange factor directly activated by cAMP RAPGEF3; cAMP-GEFI; cAMP-regulated guanine nucleotide exchange factor I; cAMP-regulated guanine nucleotide exchange factor I (cAMP-GEFI); EPAC 1; Exchange factor directly activated by cAMP 1; Exchange protein directly activated by cAMP 1; Rap guanine nucleotide exchange factor 3; Rap1 guanine-nucleotide-exchange factor directly activated by cAMP
Gene Aliases: 2310016P22Rik; 9330170P05Rik; Epac; Epac1; Rapgef3
UniProt ID: (Rat) Q9Z1C8, (Mouse) Q8VCC8
Entrez Gene ID: (Rat) 59326, (Mouse) 223864
Molecular Function:
G-protein modulator
enzyme modulator
guanyl-nucleotide exchange factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.