Novus Biologicals
Manufacturer Code:NBP288125
Catalog # NB035747
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide corresponding to a region of Mouse RAPGEF2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CNrasGEF; CNrasGEF KIAA0313 Neural RAP guanine nucleotide exchange protein nRap GEP NRAPGEP PDZ domain containing guanine nucleotide exchange factor (GEF) 1 PDZGEF1 PDZ-GEF1PDZ domain-containing guanine nucleotide exchange factor 1 RA(Ras/Rap1A-associating)-GEF RA-GEFDKFZP586O1422 Rap guanine nucleotide exchange factor (GEF) 2 rap guanine nucleotide exchange factor 2 Rap-GEP; Cyclic nucleotide ras GEF; Neural RAP guanine nucleotide exchange protein; nRap GEP; PDZ domain containing guanine nucleotide exchange factor (GEF) 1; PDZ domain-containing guanine nucleotide exchange factor 1; PDZ-GEF1; RA(Ras/Rap1A-associating)-GEF; RA-GEF-1; Rap guanine nucleotide exchange factor (GEF) 2; Rap guanine nucleotide exchange factor 2; Ras/Rap1-associating GEF-1
Gene Aliases: CNrasGEF; KIAA0313; nRap GEP; NRAPGEP; PDZ-GEF1; PDZGEF1; RA-GEF; RA-GEF-1; Rap-GEP; RAPGEF2
UniProt ID: (Human) Q9Y4G8
Entrez Gene ID: (Human) 9693
Molecular Function:
G-protein modulator
enzyme modulator
guanyl-nucleotide exchange factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.