Novus Biologicals
Manufacturer Code:NBP190812
Catalog # NBP190812
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:LSNKKIYQEEEMPESGAGSEFNRKLREEARRKKYGIVLKEFRPEDQPWLLRVNGKSGRKFKGIKKGGVTENTSYYIFTQCPDGAFEAFPVHNWYNFTPLARHRTLTAEEAEEEWERRNKVLNHFSIMQQRRLKDQDQD |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BTF4 General transcription factor IIF 74 kDa subunit general transcription factor IIF subunit 1 general transcription factor IIF polypeptide 1 (74kD subunit) general transcription factor IIF polypeptide 1 74kDa RAP74Transcription initiation factor RAP74 TF2F1 TFIIF TFIIF-alpha Transcription initiation factor IIF subunit alpha; General transcription factor IIF 74 kDa subunit; General transcription factor IIF subunit 1; general transcription factor IIF, polypeptide 1, 74kDa; TFIIF-alpha; Transcription initiation factor IIF subunit alpha; Transcription initiation factor RAP74
Gene Aliases: BTF4; GTF2F1; RAP74; TF2F1; TFIIF
UniProt ID: (Human) P35269
Entrez Gene ID: (Human) 2962
Molecular Function:
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.