Proteintech
Manufacturer Code:103771AP150UL
Catalog # 50-172-6257
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | RAP1GDS1 Fusion Protein Ag0230 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Exchange factor smgGDS; Exchange factor smgGDS GDS1 RAP1GDS1 SMG GDS protein SmgGDS; Rap1 GTPase-GDP dissociation stimulator 1; RAP1, GTP-GDP dissociation stimulator 1; SMG GDS protein; SMG P21 stimulatory GDP/GTP exchange protein
Gene Aliases: BC011279; GDS1; RAP1GDS1; SmgGDS
UniProt ID: (Human) P52306, (Mouse) E9Q912
Entrez Gene ID: (Human) 5910, (Mouse) 229877
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.