Invitrogen
Manufacturer Code:PA551662
Catalog # PIPA551662
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human RAP1GAP |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ARPP-90; Rap1 GTPase-activating protein 1; Rap1, GTPase-activating protein 1; RAP1GA1 RAP1GAP1 RAP1GAPII RAPGAP rap1 GTPase-activating protein 1; Rap1GAP
Gene Aliases: 1300019I11Rik; 2310004O14Rik; AI427470; ARPP-90; KIAA0474; RAP1GA1; RAP1GAP; RAP1GAP1; RAP1GAPII; RAPGAP
UniProt ID: (Human) P47736, (Mouse) A2ALS5
Entrez Gene ID: (Human) 5909, (Mouse) 110351
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.