Proteintech
Manufacturer Code:142291AP150UL
Catalog # 50-172-9298
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | RAP1GAP Fusion Protein Ag5469 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ARPP-90; KIAA0474 RAP1 GTPase activating protein RAP1GA1 RAP1GAP RAP1GAP1 RAP1GAPII RAPGAP; Rap1 GTPase-activating protein 1; Rap1, GTPase-activating protein 1; Rap1GAP
Gene Aliases: 1300019I11Rik; 2310004O14Rik; AI427470; ARPP-90; KIAA0474; RAP1GA1; RAP1GAP; RAP1GAP1; RAP1GAPII; RAPGAP
UniProt ID: (Human) P47736, (Mouse) A2ALS5
Entrez Gene ID: (Human) 5909, (Mouse) 110351, (Rat) 313644
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.