Invitrogen
Manufacturer Code:PA544504
Catalog # PIPA544504
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human RAP1B |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GTP-binding protein (smg p21B); GTP-binding protein smg p21B; Ras family small GTP binding protein RAP1B; RAS related protein 1b; ras-related protein rab-1B-like protein; Ras-related protein Rap-1b; Ras-related protein Rap-1b-like protein; RAS-related protein RAP1B; small GTP binding protein
Gene Aliases: 2810443E11Rik; cb1026; cb119; K-REV; OK/SW-cl.11; RAL1B; RAP1B; sb:cb119; si:dkey-31f5.4; wu:fb11c04; wu:fb74e09; wu:fk70e05
UniProt ID: (Bovine) P61223, (Human) P61224, (Rat) Q62636, (Mouse) Q99JI6, (Zebrafish) Q6TEN1
Entrez Gene ID: (Bovine) 327708, (Goat) 102174543, (Dog) 608981, (Rabbit) 100339891, (Human) 5908, (Horse) 100052025, (Rat) 171337, (Mouse) 215449, (Guinea pig) 100734094, (Zebrafish) 321107
Molecular Function:
G-protein
enzyme modulator
small GTPase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.