Invitrogen
Manufacturer Code:MA529727
Catalog # PIMA529727
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 11 |
Immunogen | Recombit Mouse LRPAP1 protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: A2MRAP A2RAP HBP44 MRAP RAP alpha-2-MRAP alpha-2-macroglobulin receptor-associated protein alpha-2-macroglobulin receptor-associated protein 1 lipoprotein receptor associated protein low density lipoprotein receptor-related protein-associated protein 1 low density lipoprotein-relate; Alpha-2-macroglobulin receptor-associated protein; Alpha-2-macroglobulin receptor-associated protein precursor (Alpha-2-MRAP) (Low density lipoprotein receptor-related protein-associated protein 1) (RAP) (Heparin binding protein-44) (HBP-44); Alpha-2-MRAP; HBP-44; Heparin-binding protein 44; low density lipoprotein receptor related protein, associated protein 1; Low density lipoprotein receptor-related protein-associated protein 1; RAP
Gene Aliases: AA617339; AI790446; AU042172; C77774; HBP44; Lrpap1; RAP
UniProt ID: (Mouse) P55302
Entrez Gene ID: (Mouse) 16976
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.