Novus Biologicals
Manufacturer Code:NB10056029
Catalog # NB10056029
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 9A725 |
Immunogen | A fusion protein containing amino acid residues 326-616 of human RANK was used as immunogen. This antibody recognizes an epitope located between amino acid residues 330-427 of human RANK (NP_003830). |
Conjugate | PE |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CD265; CD265 CD265 antigen FEO loss of heterozygosity 18 chromosomal region 1 ODFROSTS OFE OPTB7 Osteoclast differentiation factor receptor PDB2 RANKLOH18CR1 Receptor activator of NF-KB receptor activator of nuclear factor-kappa B TRANCER tumor necrosis factor receptor superfamily member 11A tumor necrosis factor receptor superfamily member 11a activator of NFKB tumor necrosis factor receptor superfamily member 11a NFKB activator; loss of heterozygosity, 18, chromosomal region 1; ODFR; Osteoclast differentiation factor receptor; Paget disease of bone 2; Receptor activator of NF-KB; receptor activator of nuclear factor-kappa B; Tumor necrosis factor receptor superfamily member 11A; tumor necrosis factor receptor superfamily member 11a, NFKB activator; tumor necrosis factor receptor superfamily, member 11a, NFKB activator
Gene Aliases: CD265; FEO; LOH18CR1; ODFR; OFE; OPTB7; OSTS; PDB2; RANK; TNFRSF11A; TRANCER
UniProt ID: (Human) Q9Y6Q6
Entrez Gene ID: (Human) 8792
Molecular Function: cytokine receptor receptor tumor necrosis factor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.