R&D Systems
Manufacturer Code:MAB6831500
Catalog # MAB6831500
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 80707 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human RANK Leu28-Gly213 Accession # Q9Y6Q6 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CD265; CD265 CD265 antigen FEO loss of heterozygosity 18 chromosomal region 1 ODFR ODFROSTS OFE OPTB7 Osteoclast differentiation factor receptor PDB2 RANKLOH18CR1 Receptor activator of NF-KB receptor activator of nuclear factor-kappa B TNFRSF11A TRANCE R TRANCER tumor necrosis factor receptor superfamily member 11A tumor necrosis factor receptor superfamily member 11a activator of NFKB tumor necrosis factor receptor superfamily member 11a NFKB activator; loss of heterozygosity, 18, chromosomal region 1; ODFR; Osteoclast differentiation factor receptor; Paget disease of bone 2; Receptor activator of NF-KB; receptor activator of nuclear factor-kappa B; Tumor necrosis factor receptor superfamily member 11A; tumor necrosis factor receptor superfamily member 11a, NFKB activator; tumor necrosis factor receptor superfamily, member 11a, NFKB activator
Gene Aliases: CD265; FEO; LOH18CR1; ODFR; OFE; OPTB7; OSTS; PDB2; RANK; TNFRSF11A; TRANCER
UniProt ID: (Human) Q9Y6Q6
Entrez Gene ID: (Human) 8792
Molecular Function:
cytokine receptor
receptor
tumor necrosis factor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.