R&D Systems
Manufacturer Code:FAB683G025
Catalog # FAB683G025
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 80704 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human RANK/TNFRSF11A extracellular domain Accession # Q9Y6Q6 |
Conjugate | Alexa Fluor 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Protect from light. Do not freeze. 12 months from date of receipt 2 to 8 degreesC as supplied. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CD265; CD265 CD265 antigen FEO loss of heterozygosity 18 chromosomal region 1 ODFR ODFROSTS OFE OPTB7 Osteoclast differentiation factor receptor PDB2 RANKLOH18CR1 Receptor activator of NF-KB receptor activator of nuclear factor-kappa B TNFRSF11A TRANCE R TRANCER tumor necrosis factor receptor superfamily member 11A tumor necrosis factor receptor superfamily member 11a activator of NFKB tumor necrosis factor receptor superfamily member 11a NFKB activator; loss of heterozygosity, 18, chromosomal region 1; ODFR; Osteoclast differentiation factor receptor; Paget disease of bone 2; Receptor activator of NF-KB; receptor activator of nuclear factor-kappa B; Tumor necrosis factor receptor superfamily member 11A; tumor necrosis factor receptor superfamily member 11a, NFKB activator; tumor necrosis factor receptor superfamily, member 11a, NFKB activator
Gene Aliases: CD265; FEO; LOH18CR1; ODFR; OFE; OPTB7; OSTS; PDB2; RANK; TNFRSF11A; TRANCER
UniProt ID: (Human) Q9Y6Q6
Entrez Gene ID: (Human) 8792
Molecular Function:
cytokine receptor
receptor
tumor necrosis factor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.