Invitrogen
Manufacturer Code:MA120302
Catalog # PIMA120302
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | LOB14-8 |
Immunogen | RANK-Fc mouse fusion protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: CD265; Ly109 ODFR OFE Rank TRANCE-R mRANK; ODFR; Osteoclast differentiation factor receptor; receptor activator of NF-kappaB; Receptor activator of NF-KB; Tumor necrosis factor receptor superfamily member 11A
Gene Aliases: Ly109; mRANK; ODFR; OFE; Rank; Tnfrsf11a; TRANCE-R
UniProt ID: (Mouse) O35305
Entrez Gene ID: (Mouse) 21934
Molecular Function: cytokine receptor receptor tumor necrosis factor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.