Invitrogen
Manufacturer Code:PA517450
Catalog # PIPA517450
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to residues near the carboxy terminus of RANK |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CD265; FEO LOH18CR1 ODFR OFE OPTB7 OSTS PDB2 RANK TRANCER TNFRSF11A; loss of heterozygosity, 18, chromosomal region 1; ODFR; Osteoclast differentiation factor receptor; Paget disease of bone 2; receptor activator of NF-kappaB; Receptor activator of NF-KB; receptor activator of nuclear factor-kappa B; Tumor necrosis factor receptor superfamily member 11A; tumor necrosis factor receptor superfamily member 11a, NFKB activator; tumor necrosis factor receptor superfamily, member 11a, NFKB activator
Gene Aliases: CD265; FEO; LOH18CR1; Ly109; mRANK; ODFR; OFE; OPTB7; OSTS; PDB2; RANK; RGD1563614; TNFRSF11A; TRANCE-R; TRANCER
UniProt ID: (Human) Q9Y6Q6, (Mouse) O35305
Entrez Gene ID: (Human) 8792, (Mouse) 21934, (Rat) 498206
Molecular Function:
cytokine receptor
receptor
tumor necrosis factor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.