Proteintech
Manufacturer Code:105521AP150UL
Catalog # 50-172-6399
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | RANGRF Fusion Protein Ag0824 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HSPC165 HSPC236 MDS5 MOG1 Ran binding protein MOG1 RANGNRF RANGRF; MOG1 homolog; Ran guanine nucleotide release factor; Ran-binding protein MOG1; RanGNRF
Gene Aliases: 2400006H24Rik; HSPC165; HSPC236; MDS5; MOG1; RANGNRF; RANGRF
UniProt ID: (Human) Q9HD47, (Mouse) Q9JIB0
Entrez Gene ID: (Human) 29098, (Mouse) 57785
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.