Proteintech
Manufacturer Code:176561AP150UL
Catalog # 50-173-1706
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | RANBP6 Fusion Protein Ag11906 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: RAN binding protein 6 RANBP6; Ran-binding protein 6; Ran-GTP binding protein; RanBP6
Gene Aliases: C630001B19; RANBP6
UniProt ID: (Human) O60518, (Mouse) Q8BIV3
Entrez Gene ID: (Human) 26953, (Mouse) 240614
Molecular Function:
G-protein modulator
enzyme modulator
transfer/carrier protein
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.